Assay Details
| Gene Symbol: c11orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016674.2 | NP_001016674.1 | 247 | 80 | ||
| GenBank mRNA | BC121515.1 | - | 243 | 80 | ||
| CR760234.2 | - | 247 | 80 | |||
More Information
Set Membership: |
|

c11orf58
Genomic Map