Assay Details
| Gene Symbol: tspyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016811.1 | NP_001016811.1 | 2 - 3 | 851 | 93 | |
| XM_012968444.2 | XP_012823898.1 | 2 - 3 | 920 | 93 | ||
| GenBank mRNA | BC167578.1 | - | 2 - 3 | 871 | 93 | |
| CR855567.1 | - | 2 - 3 | 851 | 93 | ||
More Information
Set Membership: |
|

tspyl2
Genomic Map