Assay Details
| Gene Symbol: pebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016825.2 | NP_001016825.1 | 3 - 4 | 376 | 90 | |
| GenBank mRNA | CR855528.2 | - | 3 - 4 | 376 | 90 | |
More Information
Set Membership: |
|

pebp1
Genomic Map