Assay Details
| Gene Symbol: rragc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016835.2 | NP_001016835.1 | 1 - 2 | 235 | 79 | |
| XM_012957004.2 | XP_012812458.1 | 1 - 2 | 668 | 79 | ||
| GenBank mRNA | BC123045.1 | - | 1 - 2 | 457 | 79 | |
| CR855489.2 | - | 1 - 2 | 235 | 79 | ||
More Information
Set Membership: |
|

rragc
Genomic Map