Assay Details
| Gene Symbol: aqp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016845.2 | NP_001016845.1 | 4 - 5 | 517 | 82 | |
| XM_012962938.1 | XP_012818392.1 | 5 - 6 | 563 | 82 | ||
| GenBank mRNA | BC158298.1 | - | 4 - 5 | 528 | 82 | |
| CR855446.2 | - | 4 - 5 | 517 | 82 | ||
More Information
Set Membership: |
|

aqp3
Genomic Map