Assay Details
| Gene Symbol: ywhah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016868.2 | NP_001016868.1 | 1 - 2 | 224 | 58 | |
| GenBank mRNA | BC135498.1 | - | 1 - 2 | 214 | 58 | |
| CR848634.2 | - | 1 - 2 | 224 | 58 | ||
More Information
Set Membership: |
|

ywhah
Genomic Map