Assay Details
| Gene Symbol: med7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016879.2 | NP_001016879.1 | 1 | 654 | 81 | |
| XM_012959051.2 | XP_012814505.1 | 2 | 758 | 81 | ||
| GenBank mRNA | BC091623.1 | - | 1 | 654 | 81 | |
| CR848598.2 | - | 2 | 752 | 81 | ||
More Information
Set Membership: |
|

med7
Genomic Map