Assay Details
| Gene Symbol: fer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016902.2 | NP_001016902.1 | 3 - 4 | 478 | 64 | |
| XM_012962967.2 | XP_012818421.1 | 3 - 4 | 64 | |||
| XM_012962969.2 | XP_012818423.1 | 3 - 4 | 64 | |||
| XM_012962975.2 | XP_012818429.1 | 4 - 5 | 64 | |||
| XM_012962979.2 | XP_012818433.1 | 3 - 4 | 64 | |||
| XM_018093579.1 | XP_017949068.1 | 3 - 4 | 64 | |||
| GenBank mRNA | BC168414.1 | - | 2 - 3 | 64 | ||
| CR848518.2 | - | 3 - 4 | 478 | 64 | ||
More Information
Set Membership: |
|

fer
Genomic Map