Assay Details
| Gene Symbol: oat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016936.2 | NP_001016936.1 | 8 - 9 | 1063 | 66 | |
| GenBank mRNA | BC167123.1 | - | 7 - 8 | 1026 | 66 | |
| BC167345.1 | - | 7 - 8 | 1302 | 66 | ||
| CR762076.2 | - | 8 - 9 | 1063 | 66 | ||
More Information
Set Membership: |
|

oat
Genomic Map