Assay Details
| Gene Symbol: nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016962.2 | NP_001016962.1 | 748 | 68 | ||
| GenBank mRNA | BC158422.1 | - | 763 | 68 | ||
| CR761332.2 | - | 748 | 68 | |||
More Information
Set Membership: |
|

nmral1
Genomic Map