Assay Details
| Gene Symbol: ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016988.2 | NP_001016988.1 | 4 - 5 | 202 | 64 | |
| GenBank mRNA | BC167299.1 | - | 4 - 5 | 291 | 64 | |
| CR848258.2 | - | 4 - 5 | 202 | 64 | ||
More Information
Set Membership: |
|

ufm1
Genomic Map