Assay Details
| Gene Symbol: psph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016993.3 | NP_001016993.2 | 6 - 7 | 839 | 67 | |
| XM_012957015.1 | XP_012812469.1 | 6 - 7 | 874 | 67 | ||
| XM_012957016.2 | XP_012812470.1 | 6 - 7 | 890 | 67 | ||
| GenBank mRNA | BC122888.1 | - | 6 - 7 | 839 | 67 | |
| BC158947.1 | - | 6 - 7 | 750 | 67 | ||
| CR848195.2 | - | 1 - 2 | 308 | 67 | ||
More Information
Set Membership: |
|

psph
Genomic Map