Assay Details
| Gene Symbol: hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017007.2 | NP_001017007.1 | 1 - 2 | 316 | 81 | |
| GenBank mRNA | BC090580.1 | - | 1 - 2 | 316 | 81 | |
| CR762281.2 | - | 1 - 2 | 119 | 81 | ||
More Information
Set Membership: |
|

hdgf
Genomic Map