Assay Details
| Gene Symbol: hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017007.2 | NP_001017007.1 | 5 - 6 | 788 | 98 | |
| GenBank mRNA | BC090580.1 | - | 5 - 6 | 788 | 98 | |
| CR762281.2 | - | 5 - 6 | 591 | 98 | ||
More Information
Set Membership: |
|

hdgf
Genomic Map