Assay Details
| Gene Symbol: ankrd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017037.2 | NP_001017037.1 | 2 - 3 | 283 | 76 | |
| GenBank mRNA | CR761945.2 | - | 2 - 3 | 283 | 76 | |
More Information
Set Membership: |
|

ankrd37
Genomic Map