Assay Details
| Gene Symbol: sept2-a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017039.2 | NP_001017039.1 | 3 - 4 | 219 | 123 | |
| XM_012963217.2 | XP_012818671.1 | 4 - 5 | 430 | 123 | ||
| GenBank mRNA | BC091009.1 | - | 3 - 4 | 219 | 123 | |
| CR761939.2 | - | 3 - 4 | 184 | 123 | ||
More Information
Set Membership: |
|

sept2-a
Genomic Map