Assay Details
| Gene Symbol: lfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017051.1 | NP_001017051.1 | 1 - 2 | 523 | 59 | |
| GenBank mRNA | CR761798.1 | - | 1 - 2 | 523 | 59 | |
More Information
Set Membership: |
|

lfng
Genomic Map