Assay Details
| Gene Symbol: ppib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017063.2 | NP_001017063.1 | 3 - 4 | 314 | 91 | |
| GenBank mRNA | CR761630.2 | - | 3 - 4 | 314 | 91 | |
More Information
Set Membership: |
|

ppib
Genomic Map