Assay Details
| Gene Symbol: mrps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017066.2 | NP_001017066.1 | 4 - 5 | 366 | 66 | |
| XM_012957024.2 | XP_012812478.1 | 4 - 5 | 412 | 66 | ||
| XM_018091272.1 | XP_017946761.1 | 4 - 5 | 412 | 66 | ||
| GenBank mRNA | CR761601.2 | - | 4 - 5 | 366 | 66 | |
More Information
Set Membership: |
|

mrps15
Genomic Map