Assay Details
Gene Symbol: metrnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017107.1 | NP_001017107.1 | 1 - 2 | 453 | 78 | |
GenBank mRNA | CR761246.1 | - | 1 - 2 | 453 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
