Assay Details
| Gene Symbol: alg13l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017108.2 | NP_001017108.1 | 2 - 3 | 258 | 75 | |
| GenBank mRNA | BC160420.1 | - | 2 - 3 | 266 | 75 | |
| BC168793.2 | - | 2 - 3 | 294 | 75 | ||
| CR761237.2 | - | 2 - 3 | 258 | 75 | ||
More Information
Set Membership: |
|

alg13l
Genomic Map