Assay Details
| Gene Symbol: alg13l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017108.2 | NP_001017108.1 | 3 - 4 | 404 | 100 | |
| GenBank mRNA | BC160420.1 | - | 3 - 4 | 412 | 100 | |
| BC168793.2 | - | 3 - 4 | 440 | 100 | ||
| CR761237.2 | - | 3 - 4 | 404 | 100 | ||
More Information
Set Membership: |
|

alg13l
Genomic Map