Assay Details
| Gene Symbol: tent5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017206.2 | NP_001017206.1 | 1 - 2 | 389 | 97 | |
| GenBank mRNA | CR760489.2 | - | 1 - 2 | 389 | 97 | |
More Information
Set Membership: |
|

tent5b
Genomic Map