Assay Details
| Gene Symbol: pla2g7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017267.2 | NP_001017267.1 | 5 - 6 | 528 | 92 | |
| XM_012970466.2 | XP_012825920.1 | 6 - 7 | 1103 | 92 | ||
| XM_012970467.2 | XP_012825921.1 | 6 - 7 | 711 | 92 | ||
| XM_012970468.2 | XP_012825922.1 | 6 - 7 | 871 | 92 | ||
| XM_012970469.2 | XP_012825923.1 | 6 - 7 | 681 | 92 | ||
| XM_012970470.2 | XP_012825924.1 | 5 - 6 | 592 | 92 | ||
| XM_012970471.2 | XP_012825925.1 | 5 - 6 | 498 | 92 | ||
| XM_012970472.2 | XP_012825926.1 | 5 - 6 | 767 | 92 | ||
| XM_018093914.1 | XP_017949403.1 | 6 - 7 | 1101 | 92 | ||
| GenBank mRNA | BC168792.1 | - | 5 - 6 | 536 | 92 | |
| CR855818.2 | - | 5 - 6 | 528 | 92 | ||
More Information
Set Membership: |
|

pla2g7
Genomic Map