Assay Details
| Gene Symbol: alkbh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017297.3 | NP_001017297.2 | 10 - 11 | 1453 | 72 | |
| XM_012957058.2 | XP_012812512.1 | 11 - 12 | 1568 | 72 | ||
| XM_012957059.2 | XP_012812513.1 | 11 - 12 | 1559 | 72 | ||
| XM_012957060.2 | XP_012812514.1 | 11 - 12 | 1579 | 72 | ||
| XM_012957061.2 | XP_012812515.1 | 11 - 12 | 1588 | 72 | ||
| GenBank mRNA | CR855607.2 | - | 11 - 12 | 1478 | 72 | |
More Information
Set Membership: |
|

alkbh8
Genomic Map