Assay Details
| Gene Symbol: abo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017300.2 | NP_001017300.1 | 6 - 7 | 499 | 72 | |
| XM_012968494.2 | XP_012823948.1 | 7 - 8 | 622 | 72 | ||
| XM_012968495.2 | XP_012823949.1 | 7 - 8 | 662 | 72 | ||
| XM_012968496.2 | XP_012823950.1 | 7 - 8 | 638 | 72 | ||
| XM_012968497.2 | XP_012823951.1 | 7 - 8 | 587 | 72 | ||
| XM_012968498.2 | XP_012823952.1 | 6 - 7 | 550 | 72 | ||
| GenBank mRNA | CR855597.2 | - | 6 - 7 | 499 | 72 | |
More Information
Set Membership: |
|

abo
Genomic Map