Assay Details
Gene Symbol: rsbn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017304.2 | NP_001017304.1 | 5 - 6 | 1522 | 72 | |
GenBank mRNA | CR855556.2 | - | 5 - 6 | 1522 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
