Assay Details
| Gene Symbol: ppm1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017305.2 | NP_001017305.1 | 3 - 4 | 847 | 70 | |
| GenBank mRNA | CR855546.2 | - | 3 - 4 | 847 | 70 | |
More Information
Set Membership: |
|

ppm1h
Genomic Map