Assay Details
| Gene Symbol: tbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017309.2 | NP_001017309.1 | 2 - 3 | 493 | 70 | |
| GenBank mRNA | CR855514.2 | - | 2 - 3 | 493 | 70 | |
More Information
Set Membership: |
|

tbpl2
Genomic Map