Assay Details
| Gene Symbol: hs6st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017317.1 | NP_001017317.1 | 1 - 2 | 728 | 91 | |
| GenBank mRNA | BC135520.1 | - | 1 - 2 | 893 | 91 | |
| CR761444.2 | - | 1 - 2 | 728 | 91 | ||
More Information
Set Membership: |
|

hs6st1
Genomic Map