Assay Details
| Gene Symbol: dck.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030361.1 | NP_001025532.1 | 4 - 5 | 649 | 91 | |
| GenBank mRNA | BC090567.1 | - | 4 - 5 | 649 | 91 | |
More Information
Set Membership: |
|

dck.1
Genomic Map