Assay Details
| Gene Symbol: atp12a-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030379.1 | NP_001025550.1 | 6 - 7 | 752 | 103 | |
| GenBank mRNA | BC090586.1 | - | 6 - 7 | 752 | 103 | |
| CR942688.2 | - | 6 - 7 | 733 | 103 | ||
More Information
Set Membership: |
|

atp12a-like
Genomic Map