Assay Details
| Gene Symbol: emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030386.1 | NP_001025557.1 | 3 - 4 | 320 | 82 | |
| GenBank mRNA | BC090597.1 | - | 3 - 4 | 320 | 82 | |
| CT025318.2 | - | 3 - 4 | 304 | 82 | ||
More Information
Set Membership: |
|

emp2
Genomic Map