Assay Details
| Gene Symbol: ywhaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030399.1 | NP_001025570.1 | 1 - 2 | 422 | 92 | |
| GenBank mRNA | BC090612.1 | - | 1 - 2 | 422 | 92 | |
| CT025504.2 | - | 1 - 2 | 310 | 92 | ||
More Information
Set Membership: |
|

ywhaq
Genomic Map