Assay Details
Gene Symbol: adh1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030410.1 | NP_001025581.1 | 3 - 4 | 309 | 102 | |
GenBank mRNA | BC091025.1 | - | 3 - 4 | 309 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
