Assay Details
| Gene Symbol: suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030418.1 | NP_001025589.1 | 2 - 3 | 202 | 58 | |
| GenBank mRNA | BC091038.1 | - | 2 - 3 | 202 | 58 | |
More Information
Set Membership: |
|

suclg1
Genomic Map