Assay Details
| Gene Symbol: idh3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030426.1 | NP_001025597.1 | 1 - 2 | 120 | 67 | |
| GenBank mRNA | BC091046.1 | - | 1 - 2 | 120 | 67 | |
More Information
Set Membership: |
|

idh3g
Genomic Map