Assay Details
| Gene Symbol: tha1p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030440.1 | NP_001025611.1 | 3 - 4 | 317 | 84 | |
| XM_012966670.2 | XP_012822124.1 | 3 - 4 | 392 | 84 | ||
| XM_012966671.1 | XP_012822125.1 | 3 - 4 | 497 | 84 | ||
| GenBank mRNA | BC091069.1 | - | 3 - 4 | 317 | 84 | |
More Information
Set Membership: |
|

tha1p
Genomic Map