Assay Details
| Gene Symbol: gbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030449.1 | NP_001025620.1 | 1502 | 100 | ||
| GenBank mRNA | BC091085.1 | - | 1502 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gbp4| Gene Symbol: gbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030449.1 | NP_001025620.1 | 1502 | 100 | ||
| GenBank mRNA | BC091085.1 | - | 1502 | 100 | ||
Set Membership: |
|