Assay Details
| Gene Symbol: acy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030456.1 | NP_001025627.1 | 2 - 3 | 575 | 75 | |
| XM_012957088.2 | XP_012812542.1 | 3 - 4 | 528 | 75 | ||
| GenBank mRNA | BC091576.1 | - | 2 - 3 | 575 | 75 | |
More Information
Set Membership: |
|

acy3
Genomic Map