Assay Details
| Gene Symbol: vcpkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030466.1 | NP_001025637.1 | 4 - 5 | 557 | 117 | |
| GenBank mRNA | BC091601.1 | - | 4 - 5 | 557 | 117 | |
More Information
Set Membership: |
|

vcpkmt
Genomic Map