Assay Details
| Gene Symbol: cyb5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030467.1 | NP_001025638.1 | 6 - 7 | 560 | 66 | |
| XM_012961221.2 | XP_012816675.1 | 6 - 7 | 629 | 66 | ||
| GenBank mRNA | BC091602.1 | - | 6 - 7 | 560 | 66 | |
| CR942741.2 | - | 6 - 7 | 567 | 66 | ||
More Information
Set Membership: |
|

cyb5r2
Genomic Map