Assay Details
Gene Symbol: gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030474.1 | NP_001025645.1 | 1 - 2 | 461 | 73 | |
XM_012957094.2 | XP_012812548.1 | 1 - 2 | 499 | 73 | ||
GenBank mRNA | BC091618.1 | - | 1 - 2 | 461 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
