Assay Details
| Gene Symbol: mfsd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030482.1 | NP_001025653.1 | 3 - 4 | 751 | 109 | |
| GenBank mRNA | BC091703.1 | - | 3 - 4 | 751 | 109 | |
More Information
Set Membership: |
|

mfsd4a
Genomic Map