Assay Details
| Gene Symbol: exosc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030487.1 | NP_001025658.1 | 5 - 6 | 438 | 70 | |
| GenBank mRNA | BC091582.1 | - | 5 - 6 | 438 | 70 | |
More Information
Set Membership: |
|

exosc2
Genomic Map