Assay Details
| Gene Symbol: osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030491.1 | NP_001025662.1 | 2033 | 75 | ||
| GenBank mRNA | BC077063.1 | - | 11 - 12 | 2067 | 75 | |
| BC092369.1 | - | 2033 | 75 | |||
More Information
Set Membership: |
|

osbpl11
Genomic Map