Assay Details
| Gene Symbol: scamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030501.1 | NP_001025672.1 | 989 | 65 | ||
| GenBank mRNA | BC093453.1 | - | 989 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scamp3| Gene Symbol: scamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030501.1 | NP_001025672.1 | 989 | 65 | ||
| GenBank mRNA | BC093453.1 | - | 989 | 65 | ||
Set Membership: |
|