Assay Details
| Gene Symbol: serpina3m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030522.1 | NP_001025693.1 | 3 - 4 | 816 | 94 | |
| XM_012968518.2 | XP_012823972.1 | 7 - 8 | 2130 | 94 | ||
| GenBank mRNA | BC094949.1 | - | 3 - 4 | 816 | 94 | |
More Information
Set Membership: |
|

serpina3m
Genomic Map