Assay Details
| Gene Symbol: fbxo25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030531.1 | NP_001025702.1 | 9 - 10 | 1049 | 57 | |
| GenBank mRNA | BC092557.1 | - | 9 - 10 | 1049 | 57 | |
More Information
Set Membership: |
|

fbxo25
Genomic Map