Assay Details
| Gene Symbol: vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032310.1 | NP_001027481.1 | 7 - 8 | 1673 | 59 | |
| XM_012969845.1 | XP_012825299.1 | 5 - 6 | 1277 | 59 | ||
| GenBank mRNA | BC095909.1 | - | 7 - 8 | 1673 | 59 | |
More Information
Set Membership: |
|

vangl2
Genomic Map